Merck
  • Zebularine exerts its antiproliferative activity through S phase delay and cell death in human malignant mesothelioma cells.
Zebularine exerts its antiproliferative activity through S phase delay and cell death in human malignant mesothelioma cells.
Bioscience, biotechnology, and biochemistry (2018-04-25)
Yukitoshi Takemura, Motohiko Satoh, Kenichi Hatanaka, Shunichiro Kubota
ABSTRACT
Malignant mesothelioma is an asbestos-related aggressive tumor and current therapy remains ineffective. Zebularine as a DNA methyltransferase (DNMT) inhibitor has an anti-tumor effect in several human cancer cells. The aim of the present study was to investigate whether zebularine could induce antiproliferative effect in human malignant mesothelioma cells. Zebularine induced cell growth inhibition in a dose-dependent manner. In addition, zebularine dose-dependently decreased expression of DNMT1 in all malignant mesothelioma cells tested. Cell cycle analysis indicated that zebularine induced S phase delay. Zebularine also induced cell death in malignant mesothelioma cells. In contrast, zebularine did not induce cell growth inhibition and cell death in human normal fibroblast cells. These results suggest that zebularine has a potential for the treatment of malignant mesothelioma by inhibiting cell growth and inducing cell death.
Sigma-Aldrich
Zebularine, ≥98% (HPLC), solid
Pricing and availability is not currently available.