Merck
Phytochemistry: ibuprofen-like activity in extra-virgin olive oil.
Nature (2005-09-02)
Gary K Beauchamp, Russell S J Keast, Diane Morel, Jianming Lin, Jana Pika, Qiang Han, Chi-Ho Lee, Amos B Smith, Paul A S Breslin
ABSTRACT
Newly pressed extra-virgin olive oil contains oleocanthal--a compound whose pungency induces a strong stinging sensation in the throat, not unlike that caused by solutions of the non-steroidal anti-inflammatory drug ibuprofen. We show here that this similar perception seems to be an indicator of a shared pharmacological activity, with oleocanthal acting as a natural anti-inflammatory compound that has a potency and profile strikingly similar to that of ibuprofen. Although structurally dissimilar, both these molecules inhibit the same cyclooxygenase enzymes in the prostaglandin-biosynthesis pathway.
Sigma-Aldrich
Oleocanthal, from Olea europaea