Merck
  • MicroRNA-449a Inhibition Protects H9C2 Cells Against Hypoxia/Reoxygenation-Induced Injury by Targeting the Notch-1 Signaling Pathway.
MicroRNA-449a Inhibition Protects H9C2 Cells Against Hypoxia/Reoxygenation-Induced Injury by Targeting the Notch-1 Signaling Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-05-15)
Jing Cheng, Qianfu Wu, Rong Lv, Li Huang, Banglong Xu, Xianbao Wang, Aihua Chen, Fei He
ABSTRACT
The present study aimed to detect the expression of miR-449a and investigate the effect of miR-449a on cell injury in cardiomyocytes subjected to hypoxia/ reoxygenation (H/R) and its underlying mechanisms. The expression of miR-449a was determined using reverse transcription-polymerase chain reaction in both neonatal rat ventricular myocytes and H9C2 cells. For gain-of-function and loss-of-function studies, H9C2 cells were transfected with either miR-449a mimics or miR-449a inhibitor. The target gene of miR-449a was confirmed by a dual-luciferase reporter assay. Apoptosis was analyzed by both flow cytometry using Annexin V and propidium iodide and transferase-mediated deoxyuridine triphosphate-biotin nick end labeling (TUNEL). Necrosis was confirmed by the detection of lactate dehydrogenase release. The cell viability was measured using the methylthiotetrazole method. The protein levels of Notch-1, Notch-1 intracellular domain, hairy and enhancer of split-1 (Hes-1), and apoptosis-related genes were measured by Western blot analysis. MiR-449a was significantly upregulated in both neonatal rat ventricular myocytes and H9C2 cells subjected to H/R. However, H/R-induced cell apoptosis and necrosis were markedly reduced by miR-449a inhibition. By targeting Notch-1, miR-449a regulated the Notch-1/ Hes-1 signaling pathway. The blockade of the Notch signaling pathway partly abolished the protective effect of miR-449a suppression against H/R injury, whereas the overexpression of Notch-1 intracellular domain partly reversed the effect of miR-449a overexpression on H/R-induced cell injury. The present study suggested that miR-449a inhibition protected H9C2 cells against H/R-induced cell injury by targeting the Notch-1 signaling pathway, providing a novel insight into the molecular basis of myocardial ischemia-reperfusion injury and a potential therapeutic target.
Sigma-Aldrich
5-(Methylthio)-1H-tetrazole, 97%
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%