Merck
PtdIns(4,5)P2 is not required for secretory granule docking.
Traffic (Copenhagen, Denmark) (2018-03-16)
Muhmmad Omar-Hmeadi, Nikhil R Gandasi, Sebastian Barg
ABSTRACT
Phosphoinositides (PtdIns) play important roles in exocytosis and are thought to regulate secretory granule docking by co-clustering with the SNARE protein syntaxin to form a docking receptor in the plasma membrane. Here we tested this idea by high-resolution total internal reflection imaging of EGFP-labeled PtdIns markers or syntaxin-1 at secretory granule release sites in live insulin-secreting cells. In intact cells, PtdIns markers distributed evenly across the plasma membrane with no preference for granule docking sites. In contrast, syntaxin-1 was found clustered in the plasma membrane, mostly beneath docked granules. We also observed rapid accumulation of syntaxin-1 at sites where granules arrived to dock. Acute depletion of plasma membrane phosphatidylinositol (4,5) bisphosphate (PtdIns(4,5)P2 ) by recruitment of a 5'-phosphatase strongly inhibited Ca2+ -dependent exocytosis, but had no effect on docked granules or the distribution and clustering of syntaxin-1. Cell permeabilization by α-toxin or formaldehyde-fixation caused PtdIns marker to slowly cluster, in part near docked granules. In summary, our data indicate that PtdIns(4,5)P2 accelerates granule priming, but challenge a role of PtdIns in secretory granule docking or clustering of syntaxin-1 at the release site.
Sigma-Aldrich
α-Hemolysin from Staphylococcus aureus, lyophilized powder, Protein ~60 % by Lowry, ≥10,000 units/mg protein
Pricing and availability is not currently available.