Merck
  • Thioredoxin 80-activated-monocytes (TAMs) inhibit the replication of intracellular pathogens.
Thioredoxin 80-activated-monocytes (TAMs) inhibit the replication of intracellular pathogens.
PloS one (2011-03-03)
Ximena Cortes-Bratti, Eugénie Bassères, Fabiola Herrera-Rodriguez, Silvia Botero-Kleiven, Giuseppe Coppotelli, Jens B Andersen, Maria G Masucci, Arne Holmgren, Esteban Chaves-Olarte, Teresa Frisan, Javier Avila-Cariño
ABSTRACT
Thioredoxin 80 (Trx80) is an 80 amino acid natural cleavage product of Trx, produced primarily by monocytes. Trx80 induces differentiation of human monocytes into a novel cell type, named Trx80-activated-monocytes (TAMs). In this investigation we present evidence for a role of TAMs in the control of intracellular bacterial infections. As model pathogens we have chosen Listeria monocytogenes and Brucella abortus which replicate in the cytosol and the endoplasmic reticulum respectively. Our data indicate that TAMs efficiently inhibit intracellular growth of both L. monocytogenes and B. abortus. Further analysis shows that Trx80 activation prevents the escape of GFP-tagged L. monocytogenes into the cytosol, and induces accumulation of the bacteria within the lysosomes. Inhibition of the lysosomal activity by chloroquine treatment resulted in higher replication of bacteria in TAMs compared to that observed in control cells 24 h post-infection, indicating that TAMs kill bacteria by preventing their escape from the endosomal compartments, which progress into a highly degradative phagolysosome. Our results show that Trx80 potentiates the bactericidal activities of professional phagocytes, and contributes to the first line of defense against intracellular bacteria.
Sigma-Aldrich
Guvacine hydrochloride, ≥97% (NMR), solid