4X SDS Sample Loading Buffer for Western Blotting
This SDS sample loading buffer recipe is ideal for preparing and loading protein samples into gels for polyacrylamide gel electrophoresis analysis. This recipe calculator enables the accurate preparation of a 4X SDS sample loading buffer for any volume that you need. Input your desired volume, click the CALCULATE button, and the table will populate with the amounts of each component needed.
4X Solution concentrations:
- Tris-HCl: 0.2 M
- DTT: 0.4 M
- SDS: 277 mM, 8.0% (w/v)
- Bromophenol blue: 6 mM
- Glycerol: 4.3 M
7.5 mL Recipe | Desired Volume: mL |
|
1.5 mL 1 M Tris-HCl pH 6.5 (20-160) |
mL 1 M Tris-HCl pH 6.5 |
|
3 mL of 1 M DTT (dithiothreitol) (43816) |
mL of 1 M DTT (dithiothreitol) |
|
0.6 g of SDS (sodium dodecyl sulfate) (L3771) | g of SDS (sodium dodecyl sulfate) |
|
30 mg of bromophenol blue (B0126) | mg of bromophenol blue | |
2.4 mL of glycerol (G5516) | mL of glycerol | |
Adjust final volume to 7.5 mL with Milli-Q® Water | Adjust final volume to mL with Milli-Q® Water |
|
Make 500 µL aliquots and store at -20 °C. Note: Should the solution appear orange/yellow, it may be necessary to raise the pH by adding a suitable base, such as 5 M NaOH, dropwise. |
OTHER RECIPE CALCULATORS FOR WESTERN BLOTTING BUFFERS & SOLUTIONS
Stock Solutions
- Tris-Buffered Saline (TBS) 10X Stock Solution
- 10X Phosphate-Buffered Saline (PBS)
- Stock Components for Western Blot Stripping Solutions
Wash & Blocking Buffers
- 1X Tris-Buffered Saline (TBS) Working Solution
- 1X Tris-Buffered Saline, 0.1% Tween® 20 Detergent (TBST)
- 1X PBS (Phosphate-Buffered Saline) Concentration Recipe Calculator
- 1X Phosphate-Buffered Saline, 0.1% Tween® 20 Detergent (PBST)
- Non-Fat Dry Milk (NFDM) Blocking Buffer
- Bovine Serum Albumin (BSA) Blocking Buffer Recipe
Stripping Solutions