Skip to Content
Merck
Z-N-Me-Aib-OH 98%

Z-N-Me-Aib-OH

Z-N-Me-Aib-OH 98%
Synonym(s):
Z-N,2-Dimethylalanine
Linear Formula:
C6H5CH2OCON(CH3)C(CH3)2CO2H
CAS Number:
Molecular Weight:
251.28

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing