
Zinc p-toluenesulfonate hydrate

Synonym(s):
p-Toluenesulfonic acid zinc salt
Linear Formula:
(CH3C6H4SO3)2Zn·xH2O
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing