Skip to Content
Merck
Zinc hexafluoroacetylacetonate dihydrate 98%

Zinc hexafluoroacetylacetonate dihydrate

Zinc hexafluoroacetylacetonate dihydrate 98%
Synonym(s):
Hexafluoroacetylacetone zinc derivative
Linear Formula:
Zn(C5HF6O2)2 · 2H2O
CAS Number:
Molecular Weight:
515.52

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing