
Z-Glu-OMe

Synonym(s):
N-Carbobenzyloxy-L-glutamic acid 1-methyl ester, Z-L-Glutamic acid 1-methyl ester
Linear Formula:
HOOCCH2CH2CH(NHCOOCH2C6H5)COOCH3
CAS Number:
Molecular Weight:
295.29
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing