
Z-D-Phe-OH

Synonym(s):
N-Cbz-D-phenylalanine, Z-D-phenylalanine
Empirical Formula (Hill Notation):
C17H17NO4
CAS Number:
Molecular Weight:
299.32
Products
Product Category
Brand
Formula Weight
Physical Form
Application
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing