Skip to Content
Merck
Z-Asp-OMe ≥98.0% (TLC)

Z-Asp-OMe

Z-Asp-OMe ≥98.0% (TLC)
Synonym(s):
N-Cbz-L-aspartic acid α-methyl ester, Z-L-Aspartic acid 1-methyl ester
Linear Formula:
HO2CCH2CH(NHCO2CH2C6H5)CO2CH3
CAS Number:
Molecular Weight:
281.26

Products

Brand
Formula Weight
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing