
Z-Ala-Arg-Arg 4-methoxy-β-naphthylamide acetate salt

Empirical Formula (Hill Notation):
C34H46N10O6
CAS Number:
Molecular Weight:
690.79
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing