USGS24 graphite (carbon isotopes in graphite)
Synonym(s):
Graphite
Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Isotope
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing