Skip to Content
Merck
Tris(tert-butyldimethylsilyl) phosphite ≥97.0%

Tris(tert-butyldimethylsilyl) phosphite

Tris(tert-butyldimethylsilyl) phosphite ≥97.0%
Empirical Formula (Hill Notation):
C18H45O3PSi3
CAS Number:
Molecular Weight:
424.78

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing