
Tri-sec-butylborane solution

Synonym(s):
Tris(1-methylpropyl)borane
Linear Formula:
[C2H5CH(CH3)]3B
CAS Number:
Molecular Weight:
182.15
Products
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing