![Tris(dimethylsiloxy)phenylsilane 96%](/deepweb/assets/sigmaaldrich/product/structures/258/607/6af8bec5-c8be-46d8-a4dc-e45f4137e9a5/640/6af8bec5-c8be-46d8-a4dc-e45f4137e9a5.png)
Tris(dimethylsiloxy)phenylsilane
![Tris(dimethylsiloxy)phenylsilane 96%](/deepweb/assets/sigmaaldrich/product/structures/258/607/6af8bec5-c8be-46d8-a4dc-e45f4137e9a5/640/6af8bec5-c8be-46d8-a4dc-e45f4137e9a5.png)
Synonym(s):
Phenyl-tris(dimethylsiloxy)silane
Linear Formula:
[(CH3)2SiHO]3SiC6H5
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing