![Trimethyl(phenylthiomethyl)silane 99%](/deepweb/assets/sigmaaldrich/product/structures/288/625/a2033f39-4d01-495c-84c9-ba53b4b8a121/640/a2033f39-4d01-495c-84c9-ba53b4b8a121.png)
Trimethyl(phenylthiomethyl)silane
![Trimethyl(phenylthiomethyl)silane 99%](/deepweb/assets/sigmaaldrich/product/structures/288/625/a2033f39-4d01-495c-84c9-ba53b4b8a121/640/a2033f39-4d01-495c-84c9-ba53b4b8a121.png)
Synonym(s):
(Phenylthiomethyl)trimethylsilane, (Trimethylsilyl)methyl phenyl sulfide, Phenyl (trimethylsilyl)methyl sulfide
Linear Formula:
C6H5SCH2Si(CH3)3
CAS Number:
Molecular Weight:
196.38
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing