Skip to Content
Merck
Trimethoxysilane 95%

Trimethoxysilane

Trimethoxysilane 95%
Linear Formula:
(CH3O)3SiH
CAS Number:
Molecular Weight:
122.20
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing