![Triethylmethylammonium hydroxide solution 20 wt. % in H2O](/deepweb/assets/sigmaaldrich/product/structures/256/505/6f8a0392-5f8e-4126-8f5a-72c7fb5ad8c0/640/6f8a0392-5f8e-4126-8f5a-72c7fb5ad8c0.png)
Triethylmethylammonium hydroxide solution
![Triethylmethylammonium hydroxide solution 20 wt. % in H2O](/deepweb/assets/sigmaaldrich/product/structures/256/505/6f8a0392-5f8e-4126-8f5a-72c7fb5ad8c0/640/6f8a0392-5f8e-4126-8f5a-72c7fb5ad8c0.png)
Synonym(s):
Methyltriethylammonium hydroxide
Linear Formula:
(C2H5)3N(OH)CH3
CAS Number:
Molecular Weight:
133.23
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing