Skip to Content
Merck
Triethyl 2-fluoro-2-phosphonoacetate 96%

Triethyl 2-fluoro-2-phosphonoacetate

Triethyl 2-fluoro-2-phosphonoacetate 96%
Synonym(s):
2-Fluoro-2-phosphonoacetic acid triethyl ester, Diethyl [(ethoxycarbonyl)fluoromethyl]phosphonate, Ethyl diethylphosphonofluoroacetate
Linear Formula:
(C2H5O)2P(O)CH(F)CO2C2H5
CAS Number:
Molecular Weight:
242.18

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing