
Tridecafluorohexane-1-sulfonic acid potassium salt

Synonym(s):
1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluoro-1-hexanesulfonic acid potassium salt, PFHxS potassium salt, Perfluorohexanesulfonic acid potassium salt, Potassium tridecafluoro-1-hexanesulfonate
Empirical Formula (Hill Notation):
C6F13KO3S
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing