
Trichloroacetimidate on Wang resin

Synonym(s):
4-Alkoxybenzyl trichloroacetimidate, polymer-bound, Wang trichloroacetimidate resin
Products
Brand
Cleavage Condition
Linker
Chain Type
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing