Tributylborane solution
Synonym(s):
TBB, Tributylboron
Linear Formula:
[CH3(CH2)3]3B
CAS Number:
Molecular Weight:
182.15
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing