
trans-Nonachlor solution

Empirical Formula (Hill Notation):
C10H5Cl9
Products
Product Category
Brand
Formula Weight
Product Line
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing