Skip to Content
Merck
Thulium(III) sulfate octahydrate 99.9% trace metals basis

Thulium(III) sulfate octahydrate

Thulium(III) sulfate octahydrate 99.9% trace metals basis
Synonym(s):
Sulfuric acid thulium(III) salt octahydrate
Linear Formula:
Tm2(SO4)3 · 8H2O
CAS Number:
Molecular Weight:
770.18
EC Number:

Products

Brand
Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing