Skip to Content
Merck
Tetrathiafulvalene 97%

Tetrathiafulvalene

Tetrathiafulvalene 97%
Synonym(s):
Δ2,2′-Bi-1,3-dithiole, TTF
Empirical Formula (Hill Notation):
C6H4S4
CAS Number:
Molecular Weight:
204.36
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing