Skip to Content
Merck
Tetrahydro-2-furoic acid 97%

Tetrahydro-2-furoic acid

Tetrahydro-2-furoic acid 97%
Synonym(s):
Tetrahydrofuran-2-carboxylic acid
Empirical Formula (Hill Notation):
C5H8O3
CAS Number:
Molecular Weight:
116.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing