Skip to Content
Merck
Tetrabutylammonium octachlorodirhenate(III) 98%

Tetrabutylammonium octachlorodirhenate(III)

Tetrabutylammonium octachlorodirhenate(III) 98%
Linear Formula:
{[CH3(CH2)3]4N}2Re2Cl8
CAS Number:
Molecular Weight:
1140.97

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing