Skip to Content
Merck
Tetrabutylammonium difluorotriphenylstannate 97%

Tetrabutylammonium difluorotriphenylstannate

Tetrabutylammonium difluorotriphenylstannate 97%
Synonym(s):
Tetrabutylammonium difluorotriphenyltin
Linear Formula:
(CH3CH2CH2CH2)4N[(C6H5)3SnF2]
CAS Number:
Molecular Weight:
630.48

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing