Skip to Content
Merck
tert-Amylbenzene ≥99%, H2O <100 ppm, acid <200 ppm

tert-Amylbenzene

tert-Amylbenzene ≥99%, H2O <100 ppm, acid <200 ppm
Empirical Formula (Hill Notation):
C11H16
CAS Number:
Molecular Weight:
148.24

Products

Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing