Skip to Content
Merck
Supel™ Tox Tricho volume 6 mL, pk of 30

Supel Tox Tricho

Supel™ Tox Tricho volume 6 mL, pk of 30
Synonym(s):
Supel Tox

Products

Brand
Chemical Composition
Volume (ml)
Application
Package Size
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing