
Sulfacetamide sodium salt monohydrate

Empirical Formula (Hill Notation):
C8H9N2NaO3S · H2O
CAS Number:
Molecular Weight:
254.24
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Activity Spectrum
Markush Class
Markush Group
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing