Spermine–Nitric oxide complex
Synonym(s):
N-[4-[1-(3-Aminopropyl)-2-hydroxy-2-nitrosohydrazino]butyl]-1,3-propanediamine, SPER/NO, Spermine NONOate
Empirical Formula (Hill Notation):
C10H26N6O2
CAS Number:
Molecular Weight:
262.35
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing