Skip to Content
Merck
Solvene®300/P300

Solvene®300/P300

Solvene®300/P300
Synonym(s):
Poly(vinylidene fluoride-co-trifluoroethylene)
Linear Formula:
(C2H2F2)n(C2HF3)m
CAS Number:

Products

Brand
Melting Point (°C)
Physical Form
Chemical Composition
Showing 1-1 of 1
Compare
Product Number
Description
Pricing