Skip to Content
Merck
Sodium phosphotungstate tribasic hydrate puriss. p.a.

Sodium phosphotungstate tribasic hydrate

Sodium phosphotungstate tribasic hydrate puriss. p.a.
Synonym(s):
Phosphotungstic acid trisodium salt hydrate, Tungstophosphoric acid trisodium salt
Linear Formula:
Na3[P(W3O10)4] · aq
CAS Number:
Molecular Weight:
2946.00 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing