Skip to Content
Merck
Sodium mercaptopyruvate ≥85%, technical grade

Sodium mercaptopyruvate

Sodium mercaptopyruvate ≥85%, technical grade
Synonym(s):
3-Mercapto-2-oxopropionic acid sodium salt, Mercaptopyruvic acid sodium salt
Linear Formula:
HSCH2COCOONa
CAS Number:
Molecular Weight:
142.11
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing