Skip to Content
Merck
SMDPPEH sublimed, 97% (HPLC)

SMDPPEH

SMDPPEH sublimed, 97% (HPLC)
Synonym(s):
2,5-Di-(2-ethylhexyl)-3,6-bis-(5′′-n-hexyl-[2,2′,5′,2′′]terthiophen-5-yl)-pyrrolo[3,4-c]pyrrole-1,4-dione
Empirical Formula (Hill Notation):
C58H72N2O2S6
CAS Number:
Molecular Weight:
1021.59

Products

Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing