Sinomenine
Empirical Formula (Hill Notation):
C19H23NO4 · 0.3CHCl3
CAS Number:
Molecular Weight:
365.20
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing