Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silver perchlorate hydrate
Empirical Formula (Hill Notation):
AgClO
4
· xH
2
O
CAS Number:
331717-44-3
Molecular Weight:
207.32 (anhydrous basis)
EC Number:
232-035-4
Products
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Purity
Reagent Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
226548
Silver perchlorate hydrate,
99%
Expand
View Pricing
85271
Silver perchlorate hydrate,
≥96.0% (calc. based on dry substance, AT)
Expand
View Pricing