Silicic acid
Synonym(s):
Silicic acid
Empirical Formula (Hill Notation):
O2Si · xH2O
CAS Number:
Molecular Weight:
60.08 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing