Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(
S
)-(+)-Norepinephrine L-bitartrate
Synonym(s):
(+)-α-(Aminomethyl)-3,4-dihydroxybenzyl alcohol (+)-tartrate (1:1), D-Noradrenaline L-bitartrate, D-Noradrenaline hydrogen L-tartrate, D-Norepinephrine D-bitartrate
Linear Formula:
(HO)
2
C
6
H
3
CH(CH
2
NH
2
)OH · C
4
H
6
O
6
CAS Number:
636-88-4
Molecular Weight:
319.26
EC Number:
211-268-5
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Optical Isomer
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
407453
(
S
)-(+)-Norepinephrine L-bitartrate,
99%
Expand
View Pricing