Skip to Content
Merck
(S)-(+)-2-Methoxy-2-phenylethanol 98%

(S)-(+)-2-Methoxy-2-phenylethanol

(S)-(+)-2-Methoxy-2-phenylethanol 98%
Synonym(s):
β-Methoxyphenethyl alcohol, (+)-β-Methoxyphenethyl alcohol
Linear Formula:
C6H5CH(OCH3)CH2OH
CAS Number:
Molecular Weight:
152.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing