(S)-(−)-(6,6′-Dimethoxybiphenyl-2,2′-diyl)bis(diphenylphosphine)
Synonym(s):
(S)-(−)-2,2′-Bis(diphenylphosphino)-6,6′-dimethoxy-1,1′-biphenyl, (S)-(−)-MeO-BIPHEP, SL-A101-2
Empirical Formula (Hill Notation):
C38H32O2P2
CAS Number:
Molecular Weight:
582.61
Products
Brand
Sustainability
Feature
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing