Skip to Content
Merck
(S)-4-tert-Butyl-2-oxazolidinone 99%

(S)-4-tert-Butyl-2-oxazolidinone

(S)-4-tert-Butyl-2-oxazolidinone 99%
Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing