Skip to Content
Merck
(S)-(−)-3-Benzyloxy-1,2-propanediol 99%

(S)-(−)-3-Benzyloxy-1,2-propanediol

(S)-(−)-3-Benzyloxy-1,2-propanediol 99%
Synonym(s):
(S)-Glycerol 1-benzyl ether, 1-Benzyl-sn-glycerol
Linear Formula:
C6H5CH2OCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
182.22

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing