Skip to Content
Merck
(S)-3-Amino-1,2-propanediol 98%

(S)-3-Amino-1,2-propanediol

(S)-3-Amino-1,2-propanediol 98%
Linear Formula:
H2NCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
91.11

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing