Skip to Content
Merck
(S)-(−)-2-Chloropropionic acid 99%

(S)-(−)-2-Chloropropionic acid

(S)-(−)-2-Chloropropionic acid 99%
Synonym(s):
(S)-(−)-2-Chloropropanoic acid, L-α-Chloropropionic acid
Linear Formula:
CH3CH(Cl)CO2H
CAS Number:
Molecular Weight:
108.52
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing