Skip to Content
Merck
(S)-(−)-1-Phenyl-1-butanol 97%

(S)-(−)-1-Phenyl-1-butanol

(S)-(−)-1-Phenyl-1-butanol 97%
Synonym(s):
(S)-(−)-α-Propylbenzyl alcohol
Linear Formula:
CH3CH2CH2CH(C6H5)OH
CAS Number:
Molecular Weight:
150.22

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing