
(S)-(−)-1-(4-Bromophenyl)ethylamine

Synonym(s):
(S)-(−)-4-Bromo-α-methylbenzylamine
Empirical Formula (Hill Notation):
C8H10BrN
CAS Number:
Molecular Weight:
200.08
Products
Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing